missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tenofovir
GREENER_CHOICE

Product Code. 18034322
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18034322 50 mg 50mg
18065612 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034322 Supplier Tocris Bioscience™ Supplier No. 3666/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reverse transcriptase inhibitor

Tenofovir is a selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 147127-20-6
Quantity 50 mg
Molecular Formula (R)-[[2-(6-Amino-9 H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid
Product Type Tenofovir
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 287.21
Inhibitors Tenofovir

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.