missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Terazosin Hydrochloride, ≥99%

Alpha-1 adrenoceptor antagonist

Brand:  Thermo Scientific™ J62055.MF

Product Code. 15414519

  • ONLINE EXCLUSIVE PRICE
    152.00 €
    149.00 € / 50mg
    Save 3.00 €
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mg
Powder
≥99%
C19H25N5O4·HCl
White to Off-white
Alpha-1 adrenoceptor antagonist
63074-08-8
423.90g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.