missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Terazosin hydrochloride

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
α1- and α2B-adrenoceptor antagonist (Ki values are 3.3, 0.7, 1.1, 7.7, 1510 and 78.2 nM for α1A, α1B, α1D, α2B, α2A and α2C receptors respectively). Antihypertensive following oral or intravenous administration in vivo.
Specifications
Specifications
| CAS | 63074-08-8 |
| Quantity | 50 mg |
| Molecular Formula | C19H26ClN5O4 |
| Formula Weight | 423.9 |
| Purity | >99% |
| Molecular Weight (g/mol) | 423.898 |
| Chemical Name or Material | Terazosin hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction