missing translation for 'onlineSavingsMsg'
Learn More

Terbinafine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15698725 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15698725 1 g 1g
15608735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15698725 Supplier Thermo Scientific Acros Supplier No. 461260010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78628-80-5
Molecular Formula C21H25N·HCl
Molecular Weight (g/mol) 327.89
InChI Key BWMISRWJRUSYEX-SZKNIZGXSA-N
Synonym terbinafine hydrochloride, terbinafine hcl, lamisil, terbinafine hydrochioride, lamisil krem, lamisil at, zabel, terbinafinum latin, terbinafina spanish, zimig
PubChem CID 5282481
ChEBI CHEBI:77614
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
SMILES CC(C)(C)C#CC=CCN(C)CC1=CC=CC2=CC=CC=C21.Cl

Specifications

Melting Point 214.0°C
Loss on Drying 0.5% max. (1 g, 105)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information (5% in methanol) Clear solution
Formula Weight 327.89
Percent Purity 99%
Chemical Name or Material Terbinafine hydrochloride
compliance-icons
Product Identifier
  • Terbinafine hydrochloride, 99%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Terbinafine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.