Learn More
Terbinafine hydrochloride, Tocris Bioscience™
Selective TASK3 activator
Brand:  Tocris Bioscience 6484/50
Encompass
Product Code. 15957960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.
Specifications
Specifications
78628-80-5
N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride