missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) acetate hydrate, REacton™, 99.9% (REO)

Product Code. 11312558
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312558 10 g 10g
11302558 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312558 Supplier Thermo Scientific Alfa Aesar Supplier No. 014600.09
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100587-92-6 | C6H9O6Tb | 336.06 g/mol

Terbium Acetate has important role as an activator for green phosphors used in color TV tubes. Meanwhile Terbium Acetate is also used in special lasers and as a dopant in solid-state devices. Terbium is also used in alloys and in the production of electronic devices. As a component of Terfenol-D, Terbium is of use in actuators, in naval sonar systems, sensors, in the SoundBugdevice (its first commercial application), and other magnetomechanical devices. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-92-6
Molecular Formula C6H9O6Tb
Molecular Weight (g/mol) 336.06
MDL Number MFCD00149233
InChI Key JQBILSNVGUAPMM-UHFFFAOYSA-K
IUPAC Name terbium(3+) triacetate
SMILES [Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water,moderately soluble in strong mineral acids
Formula Weight 336.08 (anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Terbium(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.