missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) nitrate hydrate, REacton™, 99.99% (REO)

Product Code. 11396868
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Pakningsstørrelse:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11396868 2 g 2g
11306878 10 g 10g
11316878 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11396868 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. 013593.04
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10043-27-3 | N3O9Tb | 344.94 g/mol

Terbium(III) nitrate hydrate is used in ceramics, glass, phosphors, lasers, and also is the important dopant for fibre amplifiers. It is also used in alloys and in the production of electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-27-3
Molecular Formula N3O9Tb
Molecular Weight (g/mol) 344.94
MDL Number MFCD00011257
InChI Key YJVUGDIORBKPLC-UHFFFAOYSA-N
IUPAC Name terbium(3+) trinitrate
SMILES [Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 2 g
UN Number UN1477
Merck Index 14,9157
Solubility Information Soluble in water.
Formula Weight 344.94 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Assay (REO)
Chemical Name or Material Terbium(III) nitrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.