missing translation for 'onlineSavingsMsg'
Learn More

Terbium ingot, 99.9% (REO), Ta <0.4%

Product Code. 11396517
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396517 10 g 10g
11306527 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396517 Supplier Thermo Scientific Alfa Aesar Supplier No. 000320.09
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Ingot
Quantity 10 g
Assay Percent Range 99.9% (REO)
Packaging Air-stable poly bottle
Appearance Silver Gray ingot
Density 8.27g/cm3
Chemical Name or Material Terbium, Rare Earth Oxides Content, Ta <0.2%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.