missing translation for 'onlineSavingsMsg'
Learn More

Terbufos, TRC

Product Code. 30422514
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422514 500 mg -
30409277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422514 Supplier Toronto Research Chemicals Supplier No. TRCT109000500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13071-79-9
Molecular Formula C9 H21 O2 P S3
Molecular Weight (g/mol) 288.43
Synonym Phosphorodithioic acid, S-[(tert-butylthio)methyl] O,O-diethyl ester (6CI,7CI,8CI), Methanethiol, (tert-butylthio)-, S-ester with O,O-diethyl phosphorodithioate (8CI), AC 92100, Counter, Counter 15G, Cyanater, O,O-Diethyl S-(tert-butylthio)methyl phosphorodithioate, O,O-Diethyl S-[[(1,1-dimethylethyl)thio]methyl] phosphorodithioate, S-[(tert-Butylthio)methyl] O,O-diethyl phosphorodithioate, Terbufos, Terbuphos, Terbutifos
IUPAC Name tert-butylsulfanylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)SCSC(C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H21O2PS3/c1-6-10-12(13,11-7-2)15-8-14-9(3,4)5/h6-8H2,1-5H3
Formula Weight 288.0441
Chemical Name or Material Terbufos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.