missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic dihydrazide, 95%

Product Code. 11314424
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314424 25 g 25g
11304424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314424 Supplier Thermo Scientific Alfa Aesar Supplier No. L01017.14
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136-64-1 | C8H10N4O2 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-64-1
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014762
InChI Key ALHNLFMSAXZKRC-UHFFFAOYSA-N
Synonym terephthalic dihydrazide, terephthalohydrazide, terephthalic acid dihydrazide, terephthalic hydrazide, 1,4-benzenedicarboxylic acid, dihydrazide, terephthaloyl dihydrazide, terephthalic acid bishydrazide, terephthalic acid, dihydrazide, terephthalhydrazide, terephthalicdihydrazide
PubChem CID 67294
IUPAC Name benzene-1,4-dicarbohydrazide
SMILES NNC(=O)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 982939
Formula Weight 194.19
Percent Purity 95%
Chemical Name or Material Terephthalic dihydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.