missing translation for 'onlineSavingsMsg'
Learn More

Terpin hydrate, MedChemExpress

Product Code. 16442309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442309 Supplier Medchem Express Supplier No. HYB1063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terpin hydrate is an expectorant, commonly used to loosen mucus in patients presenting with acute or chronic bronchitis, and related conditions.

Terpin hydrate is an expectorant, commonly used to loosen mucus in patients presenting with acute or chronic bronchitis, and related conditions.

Specifications

Chemical Name or Material Terpin hydrate
CAS 2451-01-6
Color White
Purity Grade Notes Research
Molecular Formula C10H22O3
Quantity 1 g
Synonym Terpin monohydrate cis-Terpin hydrate
Solubility Information DMSO : ≥ 100 mg/mL (525.54 mM) ∣H2O : 1 mg/mL (5.26 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@]1(C)CC[C@H](C(C)(C)O)CC1.O
Molecular Weight (g/mol) 190.28
Formula Weight 190.28
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.