missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol

Product Code. 10752631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752631 50 g Glass bottle 50g
10656422 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752631 Supplier Thermo Scientific Acros Supplier No. 203740500
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-55-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-55-5 , 586-81-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 18°C
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 214°C to 224°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range gamma-terpineol: balance (GC)
Packaging Glass bottle
Refractive Index 1.4820 to 1.485
Quantity 50 g
Beilstein 06, II, 66
Merck Index 15, 9313
Specific Gravity 0.933
Solubility Information Solubility in water: negligible. Other solubilities: soluble in organic solvents ,diethyl ether,,ethanol,benzene,propylene glycol
Formula Weight 154.25
Percent Purity 97%
Concentration or Composition (by Analyte or Components) alpha-terpineol: 85 to 90%, gamma-terpineol: balance (GC)
Physical Form Liquid
Chemical Name or Material α-Terpineol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Terpineol, mixture of alpha- and gamma- terpineol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.