missing translation for 'onlineSavingsMsg'
Learn More

Terpineol, Pure, Mixed Isomers, Fisher Chemical

Product Code. 10102500 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10102500 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10102500 Supplier Fisher Chemical Supplier No. T/0420/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 8000-41-7
Melting Point 31°C
Color White
Boiling Point 214°C
Packaging Amber glass bottle
Molecular Formula C10H18O
MDL Number MFCD00001557,MFCD00166983
Quantity 500 mL
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
InChI Key WUOACPNHFRMFPN-UHFFFAOYNA-N
SMILES CC1=CCC(CC1)C(C)(C)O
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Molecular Weight (g/mol) 154.25
PubChem CID 17100
ChEBI CHEBI:22469
Formula Weight 154.125g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Terpineol
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.