missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth, Modified (Granulated)

Product Code. 11395992
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Poly Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11395992 2 kg Poly Bottle 2kg
11306002 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395992 Supplier Fisher BioReagents™ Supplier No. BP97282
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 47.6g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4 mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Greater peptone and yeast concentration allow for higher plasmid yield
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
Formulation Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.