missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanethiol, TRC

Product Code. 30340545
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340545 1 L 1 L
30342990 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30340545 Supplier Toronto Research Chemicals Supplier No. TRCB6897251L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert.-Butanethiol
CAS 75-66-1
Molecular Formula C4 H10 S
Quantity 1 L
Synonym 2-Propanethiol, 2-methyl- (8CI, 9CI, ACI), 2-Methyl-2-propanethiol (ACI), 1,1-Dimethylethanethiol, 2-Isobutanethiol, t-Butanethiol, t-Butylmercaptan, t-Butylthiol, TBM, tert-Butanethiol, tert-Butyl mercaptan, tert-Butylthiol
InChI Formula InChI=1S/C4H10S/c1-4(2,3)5/h5H,1-3H3
SMILES CC(C)(C)S
IUPAC Name 2-methylpropane-2-thiol
Molecular Weight (g/mol) 90.19
Formula Weight 90.0503
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.