missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butanol, Honeywell™

Product Code. 15622510 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15622510 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15622510 Supplier Honeywell Research Chemicals Supplier No. 241271L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-65-0
Density 0.775g/mL at 25°C
Quantity 1 L
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Molecular Formula C4H10O
Linear Formula (CH3)3COH
Vapor Pressure 31mmHg (20°C), 44mmHg (26°C)
MDL Number MFCD00004464
UN Number 1120
Beilstein 906698
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILES CC(C)(C)O
IUPAC Name 2-methylpropan-2-ol
Molecular Weight (g/mol) 74.123
PubChem CID 6386
ChEBI CHEBI:45895
Formula Weight 74.12g/mol
Percent Purity ≥99% (GC)
Chemical Name or Material Tert-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.