missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, Puriss. p.a., ACS reagent, ≥99.7% (GC), Honeywell Riedel-de Haen™

Product Code. 16685753
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685753 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685753 Supplier Honeywell Riedel-de Haën Supplier No. 1946025KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-65-0
Color Colorless
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Quantity 25 kg
Flash Point 15°C
Molecular Formula C4H10O
Linear Formula (CH3)3COH
Beilstein 906698
Synonym 2-Methyl-2-propanol; tert-Butyl alcohol; Trimethyl carbinol
Boiling Point 81°C to 83°C (1013 hPa)
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILES CC(C)(C)O
IUPAC Name 2-methylpropan-2-ol
Molecular Weight (g/mol) 74.12
Grade ACS
Chemical Name or Material tert-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.