missing translation for 'onlineSavingsMsg'
Learn More

tert-Butoxycarbonyl-epsilon-aminocaproic Acid, TRC

Product Code. 30335642
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335642 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335642 Supplier Toronto Research Chemicals Supplier No. TRCB6906555G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butoxycarbonyl-epsilon-aminocaproic Acid
CAS 6404-29-1
Molecular Formula C11H21NO4
Quantity 5 g
Synonym 6-[N-(tert-Butoxycarbonyl)amino]caproic Acid, BOC-epsilon-aminocaproic Acid, Boc-6-aminohexanoic Acid, N-(tert-Butoxycarbonyl)-6-aminocaproic Acid, N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid, N-(tert-Butoxycarbonyl)-epsilon-aminocaproic Acid, N-(tert-Butoxycarbonyl)-epsilon-aminohexanoic Acid, N-(tert-Butyloxycarbonyl)-epsilon-aminocaproic Acid
InChI Formula InChI=1S/C11H21NO4/c1-11(2,3)16-10(15)12-8-6-4-5-7-9(13)14/h4-8H2,1-3H3,(H,12,15)(H,13,14)
SMILES CC(C)(C)OC(=O)NCCCCCC(=O)O
IUPAC Name 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Molecular Weight (g/mol) 231.29
Formula Weight 231.1471
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.