missing translation for 'onlineSavingsMsg'
Learn More

tert-Butoxytitanium (IV), TRC

Product Code. 30452756
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452756 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452756 Supplier Toronto Research Chemicals Supplier No. TRCT41447010ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butoxytitanium (IV)
CAS 3087-39-6
Molecular Formula 4(C4H10O) . Ti
Quantity 10 mL
Synonym tert-Butyl titanate(IV), tert-Butyl titanate(IV) (7CI), Orgatix TA 80, Tetra-tert-butoxytitanium, Tetra-tert-butyl titanate, Tetrakis(tert-butanolato)titanium, Tetrakis(tert-butoxy)titanium, Tetratertbutyl Orthotitanate, Titanium Tert-butoxide, Titanium Tert-butylate, Titanium Tetra-tert-butoxide, Titanium Tetrakis(2-methyl-2-propoxide), Titanium Tetrakis(t-butoxide), Titanium Tetrakis(tert-butanolate), Titanium Tetrakis(tert-butoxide), Titanium(4+) tert-Butoxide, Titanium(IV) tert-Butoxide, t-Butanol Titanium(4+) Salt, tert-Butoxytitanium (IV), tert-Butyl Orthotitanate, tert-Butyl Titanate
InChI Formula InChI=1S/4C4H9O.Ti/c4*1-4(2,3)5;/h4*1-3H3;/q4*-1;+4
SMILES CC(C)(C)O[Ti](OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C
IUPAC Name tetrakis[(2-methylpropan-2-yl)oxy]titanium
Molecular Weight (g/mol) 340.32
Formula Weight 340.2093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.