missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2,2,2-trichloroacetimidate, TRC

Product Code. 30337861
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337861 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337861 Supplier Toronto Research Chemicals Supplier No. TRCB80788025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl 2,2,2-trichloroacetimidate
CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Quantity 25 g
Synonym 2,2,2-Trichloroethanimidic acid tert-butyl ester, O-tert-Butyl trichloroacetimidate, tert-Butyl 2,2,2-trichloroacetimidate, tert-Butyl trichloroacetimidate
InChI Formula InChI=1S/C6H10Cl3NO/c1-5(2,3)11-4(10)6(7,8)9/h10H,1-3H3
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol) 218.51
Formula Weight 216.9828
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.