missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (2-bromothien-3-yl)methylcarbamate, 95%, Thermo Scientific™

Product Code. 10746925
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10746925 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10746925 Supplier Thermo Scientific Maybridge Supplier No. CC56642DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 910036-94-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H14BrNO2S
MDL Number MFCD09025882
Quantity 1 g
Synonym tert-butyl 2-bromothien-3-yl methylcarbamate, tert-butyl n-2-bromothiophen-3-yl methyl carbamate, tert-butyl 2-bromothiophen-3-yl methyl carbamate, tert-butyl 2-bromothiophen-3-yl methylcarbamate
InChI Key RFNWODFBZMWIEH-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCC1=C(SC=C1)Br
IUPAC Name tert-butyl N-[(2-bromothiophen-3-yl)methyl]carbamate
Molecular Weight (g/mol) 292.191
PubChem CID 24229570
Formula Weight 292.2
Percent Purity 95%
Chemical Name or Material tert-Butyl (2-bromothien-3-yl)methylcarbamate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.