missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-chloro-1,3-thiazole-5-carboxylate, 95%, Thermo Scientific™

Product Code. 10215374
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215374 1 g Amber Glass Bottle 1g
10434745 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215374 Supplier Thermo Scientific Maybridge Supplier No. MO08181DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-60-2
Molecular Formula C8H10ClNO2S
Molecular Weight (g/mol) 219.683
MDL Number MFCD09879991
InChI Key SQZXVWDARSIXTM-UHFFFAOYSA-N
Synonym tert-butyl 2-chlorothiazole-5-carboxylate, t-butyl-2-chloro-1,3-thiazole-5-carboxylate
PubChem CID 24229786
IUPAC Name tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
SMILES CC(C)(C)OC(=O)C1=CN=C(S1)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.69
Percent Purity 95%
Chemical Name or Material tert-Butyl 2-chloro-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.