missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3-aminopropyl)carbamate, 97%, BLD Pharm™

Product Code. 31447837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31447837 5 g
31438750 10 g
31441978 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31447837 Supplier BLD Pharm Supplier No. BD107665g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75178-96-0
Molecular Formula C8H18N2O2
MDL Number MFCD00210021
Quantity 5 g
SMILES NCCCNC(OC(C)(C)C)=O
Molecular Weight (g/mol) 174.24
Percent Purity 97%
Chemical Name or Material tert-Butyl (3-aminopropyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.