missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-Aminopropyl(methyl) Carbamate, TRC

Product Code. 30348441
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348441 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348441 Supplier Toronto Research Chemicals Supplier No. TRCB807045250MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl 3-Aminopropyl(methyl) Carbamate
CAS 150349-36-3
Molecular Formula C9H20N2O2
Quantity 250 mg
Synonym 2-Methylpropan-2-yl N-(3-aminopropyl)-N-methylcarbamate, 3-(N-tert-Butoxycarbonyl-N-methylamino)propylamine, N-(3-Aminopropyl)-N-(methyl)carbamic Acid tert-butyl Ester, N-(3-Aminopropyl)-N-methylcarbamic Acid 1,1-dimethylethyl Ester, tert-Butyl (3-aminopropyl)(methyl)carbamate
InChI Formula InChI=1S/C9H20N2O2/c1-9(2,3)13-8(12)11(4)7-5-6-10/h5-7,10H2,1-4H3
SMILES CN(CCCN)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Molecular Weight (g/mol) 188.27
Formula Weight 188.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.