missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 3-endo-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate, Thermo Scientific™

Product Code. 10380134
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10380134 1 g Amber Glass Bottle 1g
11396845 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10380134 Supplier Thermo Scientific Maybridge Supplier No. CC74315DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 143557-91-9
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD09966165
InChI Key SEGZJJSZYOEABC-IDKOKCKLSA-N
PubChem CID 58414652
IUPAC Name tert-butyl (5S)-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1CC(C2)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material Tert-butyl 3-endo-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.