missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™

Product Code. 10074962
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10074962 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10074962 Supplier Thermo Scientific Maybridge Supplier No. MO07069DA
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 690632-14-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C16H24N2O3
MDL Number MFCD05865139
Quantity 1 g
Synonym tert-butyl 4-2-aminophenoxy piperidine-1-carboxylate, tert-butyl 4-2-aminophenoxy tetrahydro-1 2h-pyridinecarboxylate, 1-boc-4-2-aminophenoxy-piperidine, tert-butyl4-2-aminophenoxy piperidine-1-carboxylate, tert-butyl 4-2-aminophenoxy tetrahydro-1 2h pyridinecarboxylate, tert-butyl4-2-aminophenoxy tetrahydro-1 2h-pyridinecarboxylate
InChI Key ISHIDQUFPVMERM-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=CC=C2N
IUPAC Name tert-butyl 4-(2-aminophenoxy)piperidine-1-carboxylate
Molecular Weight (g/mol) 292.379
PubChem CID 2794768
Formula Weight 292.38
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(2-aminophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.