missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-formylbenzyl)tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10164642
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164642 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164642 Supplier Thermo Scientific Maybridge Supplier No. CC30404DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 850375-08-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C17H24N2O3
MDL Number MFCD06658977
Quantity 1 g
Synonym tert-butyl 4-3-formylbenzyl piperazine-1-carboxylate, tert-butyl 4-3-formylbenzyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-3-formylphenyl methyl piperazine-1-carboxylate, tert-butyl4-3-formylbenzyl piperazine-1-carboxylate, 4-3-formylbenzyl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-3-formylphenyl methyl-, 1,1-dimethylethyl ester, 4-3-formylbenzyl piperazine, 3-1-piperidinyl benzaldehyde, n4-boc protected, 1-3-formyl-phenyl-piperazine-4-carboxylic acid t, 3-piperazin-4-yl methyl benzaldehyde, n1-boc protected
InChI Key YIKZZJRCKPEHGU-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=CC(=C2)C=O
IUPAC Name tert-butyl 4-[(3-formylphenyl)methyl]piperazine-1-carboxylate
Molecular Weight (g/mol) 304.39
PubChem CID 2795522
Formula Weight 304.39
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(3-formylbenzyl)tetrahydro-1(2H)-pyrazinecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.