missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™

Product Code. 10692653
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10692653 250 mg Amber Glass Bottle 250mg
10325352 1 g Amber Glass Bottle 1g
10492824 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10692653 Supplier Thermo Scientific Maybridge Supplier No. SPB08457CB
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 156185-63-6
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
InChI Key OXPWHPCCUXESFQ-UHFFFAOYSA-N
Synonym tert-butyl 4-3-hydroxypropyl piperidine-1-carboxylate, tert-butyl 4-3-hydroxypropyl tetrahydro-1 2h-pyridinecarboxylate, 3-1-boc-4-piperidyl-1-propanol, 1-n-boc-4-3'-propanol-piperdine, 3-n-boc-piperidin-4-yl propanol, boc-4-piperidinepropanol, 4-3-hydroxypropyl piperidine, n-boc protected, 1-n-boc-4-3'-propanol-piperidine, 4-3-hydroxy-propyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-3-hydroxypropyl piperidinecarboxylate
PubChem CID 2800739
IUPAC Name tert-butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)CCCO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 243.34
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.