missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-[4-(aminomethyl)benzyloxy]piperidine-1-carboxylate, 90%, Thermo Scientific™

Product Code. 10727125
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10727125 5 g Amber Glass Bottle 5g
10062224 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10727125 Supplier Thermo Scientific Maybridge Supplier No. CC67913DE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 946409-35-4
Molecular Formula C18H28N2O3
Molecular Weight (g/mol) 320.433
MDL Number MFCD12198114
InChI Key NZWAUTDCSSMVGG-UHFFFAOYSA-N
Synonym tert-butyl 4-4-aminomethyl benzyloxy piperidine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl methoxy piperidine-1-carboxylate
PubChem CID 43811047
IUPAC Name tert-butyl 4-[[4-(aminomethyl)phenyl]methoxy]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 320.43
Percent Purity 90%
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)benzyloxy]piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.