missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate, 90%, Thermo Scientific™

Product Code. 10632895
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632895 5 g Amber Glass Bottle 5g
10622895 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10632895 Supplier Thermo Scientific Maybridge Supplier No. CC67904DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1080028-74-5
Molecular Formula C18H25NO4
Molecular Weight (g/mol) 319.401
MDL Number MFCD11841070
InChI Key WQFHJESQGSVMDW-UHFFFAOYSA-N
Synonym tert-butyl 4-4-formylbenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-formylphenyl methoxy piperidine-1-carboxylate, 1-4-formylbenzyl oxy piperidine, n-boc protected, tert-butyl 4-4-formylbenzyl oxy piperidine-1-carboxylate, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzaldehyde
PubChem CID 33589526
IUPAC Name tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C=O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 319.4
Percent Purity 90%
Chemical Name or Material Tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.