missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate, ≥90%, Thermo Scientific™

Product Code. 10431703
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431703 1 g Amber Glass Bottle 1g
10066162 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431703 Supplier Thermo Scientific Maybridge Supplier No. CC30046DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 681508-91-8
Molecular Formula C17H27N3O2
Molecular Weight (g/mol) 305.422
MDL Number MFCD08060503
InChI Key VERAVZPFPZKVCT-UHFFFAOYSA-N
Synonym tert-butyl 4-4-methylamino methyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-methylaminomethyl phenyl piperazine-1-carboxylate
PubChem CID 7164564
IUPAC Name tert-butyl 4-[4-(methylaminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CNC

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 305.42
Percent Purity 90%
Chemical Name or Material Tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.