missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 11300627
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11300627 250 mg Amber Glass Bottle 250mg
10172894 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11300627 Supplier Thermo Scientific Maybridge Supplier No. CC43904CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 479226-10-3
Molecular Formula C15H21N3O3
Molecular Weight (g/mol) 291.351
MDL Number MFCD08271923
InChI Key NETKWTAEPIGRLQ-UHFFFAOYSA-N
PubChem CID 18525767
IUPAC Name tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester
Formula Weight 291.35
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.