missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate, 95+%, Thermo Scientific™

Product Code. 11386965
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386965 25 g Amber Glass Bottle 25g
10426302 1 g Amber Glass Bottle 1g
10601133 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386965 Supplier Thermo Scientific Maybridge Supplier No. TL00952EB
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 214834-18-1
Molecular Formula C11H20N2O2S
Molecular Weight (g/mol) 244.353
InChI Key SCGQNJHAAYUQOO-UHFFFAOYSA-N
Synonym 1-boc-4-thiocarbamoylpiperidine, tert-butyl 4-aminocarbothioyl tetrahydropyridine-1 2h-carboxylate, 1-tert-butoxycarbonylpiperidine-4-carbothioamide, tert-butyl 4-thiocarbamoylpiperidine-1-carboxylate, tert-butyl 4-aminocarbonothioyl piperidine-1-carboxylate, 4-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, piperidine-4-thiocarboxamide, n1-boc protected, 1-tert-butoxycarbonyl-4-carbamothioylpiperidine, 1-tert-butoxycarbonyl piperidine-4-thiocarboxamide, tert-butyl 4-thiocarbamoyl-piperidine-1-carboxylate
PubChem CID 2735648
IUPAC Name tert-butyl 4-carbamothioylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=S)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.