missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate, 97%

Product Code. 11966341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11966341 1 g 1g
11976341 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966341 Supplier Thermo Scientific Acros Supplier No. 430990010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 782501-25-1 | C10H18ClNO4S | 283.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 782501-25-1
Molecular Formula C10H18ClNO4S
Molecular Weight (g/mol) 283.77
MDL Number MFCD01861750
InChI Key VJAHMDQRVLEOFG-UHFFFAOYSA-N
Synonym 1-boc-4-chlorosulfonylpiperidine, tert-butyl 4-chlorosulfonyl piperidine-1-carboxylate, 4-chlorosulfonylpiperidine-1-carboxylic acid tert-butyl ester, 1-boc-4-piperidinesulfonyl chloride, 4-chlorosulfonyl-piperidine-1-carboxylic acid tert-butyl ester, 1-boc-piperidine-4-sulfonyl chloride, 1-boc-4-piperidinesulfonylchloride, n-boc-piperidine-4-sulfonyl chloride, tert-butyl 4-chlorosulfonyl piperidine-1-carboxyl, tert-butyl 4-chlorosulphonyl piperidine-1-carboxylate
PubChem CID 45789737
SMILES CC(C)(C)OC(=O)N1CCC(CC1)S(Cl)(=O)=O

Specifications

Melting Point 82.0°C to 85.0°C
Color White
Elemental Analysis 40.84 to 43.81% C, 4.76 to 5.11% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
UN Number 3265
IUPAC Name tert-butyl 4-(chlorosulfonyl)piperidine-1-carboxylate
Formula Weight 283.77
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material tert-Butyl 4-(Chlorosulfonyl)piperidine-1-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl 4-(chlorosulfonyl)piperidine-1-carboxylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.