missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acetoacetate, 97%

Product Code. 10366120 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
missing translation for 'unitSize'
100mL
500mL
This item is not returnable. View return policy

Product Code. 10366120

missing translation for 'mfr': Thermo Scientific 107781000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1694-31-1 | C8H14O3 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
MDL Number MFCD00008811
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -38°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 190°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CO2C(CH3)3
Refractive Index 1.4190 to 1.421
Quantity 100 mL
Fieser 01,83
Specific Gravity 0.97
Solubility Information Solubility in water: 9g/L water (20°C). Other solubilities: soluble in xylene,miscible with most organic solvents
Formula Weight 158.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl acetoacetate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.