missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acetoacetate, 97%

Product Code. 11449003
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449003 500 g 500g
11439003 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449003 Supplier Thermo Scientific Alfa Aesar Supplier No. A11115.36
 Restricted Item
In Stock
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolitik

CAS: 1694-31-1 | C8H14O3 | 158.197 g/mol

tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ?,β - unsaturated ketones synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.

Solubility
Soluble in water (9g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008811
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C

Tekniske data

Melting Point -38°C
Density 0.961
pH 5.7
Boiling Point 71°C to 72°C (11 mmHg)
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.42
Quantity 500 g
Beilstein 1680303
Solubility Information Soluble in water (9g/L).
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material tert-Butyl acetoacetate
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.