missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butyl Alcohol, Extra Pure, SLR, Fisher Chemical™

Product Code. 10611854 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
HDPE Bottle
Plastic Container
Unit Size:
2.5 L
25 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10611854 25 L Plastic Container 25 L
10458400 500 mL HDPE Bottle 500mL
10121710 2.5 L HDPE Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10611854 Supplier Fisher Chemical Supplier No. B/5250/25
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number 4464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

pH 7
Melting Point 25°C
Quantity 25 L
Packaging Plastic Container
Physical Form Liquid
Vapor Pressure 36mbar at 20°C
Viscosity 6.43 mPaS (25°C)
Boiling Point 83°C
Formula Weight 74.12g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.