missing translation for 'onlineSavingsMsg'
Learn More

Tert-butyl Alcohol, TRC

Product Code. 30410379
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410379 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410379 Supplier Toronto Research Chemicals Supplier No. TRCT11754025ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material tert.-Butanol
CAS 75-65-0
Molecular Formula C4 H10 O
Quantity 25 mL
Synonym tert-Butyl alcohol (8CI), 2-Methyl-2-propanol, 1,1-Dimethylethanol, 2-Hydroxy-2-methylpropane, Trimethylcarbinol, Trimethylmethanol, t-Butanol, t-Butanol, tert-Butanol
InChI Formula InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3
SMILES CC(C)(C)O
IUPAC Name 2-methylpropan-2-ol
Molecular Weight (g/mol) 74.12
Formula Weight 74.0732
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.