missing translation for 'onlineSavingsMsg'
Learn More

Tert-butyl Alcohol, TRC

Product Code. 30415476
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415476 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415476 Supplier Toronto Research Chemicals Supplier No. TRCT1175405ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert.-Butanol
CAS 75-65-0
Molecular Formula C4 H10 O
Quantity 5 mL
Synonym tert-Butyl alcohol (8CI), 2-Methyl-2-propanol, 1,1-Dimethylethanol, 2-Hydroxy-2-methylpropane, Trimethylcarbinol, Trimethylmethanol, t-Butanol, t-Butanol, tert-Butanol
InChI Formula InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3
SMILES CC(C)(C)O
IUPAC Name 2-methylpropan-2-ol
Molecular Weight (g/mol) 74.12
Formula Weight 74.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.