missing translation for 'onlineSavingsMsg'
Learn More

Tert-butyl Alcohol, TRC

Product Code. 30422667
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity unitSize
30422667 50 mL 50mL
1 options
This item is not returnable. View return policy

Product Code. 30422667

Brand: Toronto Research Chemicals TRCT11754050ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert.-Butanol
CAS 75-65-0
Molecular Formula C4 H10 O
Quantity 50 mL
Synonym tert-Butyl alcohol (8CI), 2-Methyl-2-propanol, 1,1-Dimethylethanol, 2-Hydroxy-2-methylpropane, Trimethylcarbinol, Trimethylmethanol, t-Butanol, t-Butanol, tert-Butanol
InChI Formula InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3
SMILES CC(C)(C)O
IUPAC Name 2-methylpropan-2-ol
Molecular Weight (g/mol) 74.12
Formula Weight 74.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.