missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Carbamate, TRC

Product Code. 30332713
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30332713 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30332713

Brand: Toronto Research Chemicals TRCB6928605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Carbamate
CAS 4248-19-5
Molecular Formula C5H11NO2
Quantity 5 g
Synonym Carbamic Acid tert-Butyl Ester, NSC 131089, O-tert-Butyl Carbamate, Boc-ammonia
InChI Formula InChI=1S/C5H11NO2/c1-5(2,3)8-4(6)7/h1-3H3,(H2,6,7)
SMILES CC(C)(C)OC(=O)N
IUPAC Name tert-butyl carbamate
Molecular Weight (g/mol) 117.15
Formula Weight 117.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.