missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloroacetate, 97%

Product Code. 10753524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753524 500 g Glass bottle 500g
10356451 25 g Glass bottle 25g
10254751 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10753524 Supplier Thermo Scientific Acros Supplier No. 347235000
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-59-5 | C6H11ClO2 | 150 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Color Colorless to Yellow
Density 1.0530g/mL
Boiling Point 48.0°C to 49.0°C (11.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CO2C(CH3)3
Refractive Index 1.4220 to 1.4240
Quantity 500 g
Beilstein 02, III, 444
Fieser 01,86; 03,36
Merck Index 15, 1564
Specific Gravity 1.053
Solubility Information Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in isopropanol,1,2-dichloroethane,toluene
Formula Weight 150
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl chloroacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.