missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Cyanoacetate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl Cyanoacetate |
| CAS | 1116-98-9 |
| Molecular Formula | C7H11NO2 |
| Quantity | 10 g |
| Synonym | Cyanoacetic Acid 1,1-Dimethylethyl Ester, Cyanoacetic Acid tert-Butyl Ester, 1,1-Dimethylethyl Cyanoacetate, Cyanoacetic Acid tert-Butyl Ester, NSC 1072, tert-Butyl 2-Cyanoacetate, tert-Butyl Cyanoacetate |
| InChI Formula | InChI=1S/C7H11NO2/c1-7(2,3)10-6(9)4-5-8/h4H2,1-3H3 |
| SMILES | CC(C)(C)OC(=O)CC#N |
| IUPAC Name | tert-butyl 2-cyanoacetate |
| Molecular Weight (g/mol) | 141.17 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?