missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Cyanoacetate, TRC

Product Code. 30341309
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341309 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341309 Supplier Toronto Research Chemicals Supplier No. TRCB6940055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Cyanoacetate
CAS 1116-98-9
Molecular Formula C7H11NO2
Quantity 5 g
Synonym Cyanoacetic Acid 1,1-Dimethylethyl Ester, Cyanoacetic Acid tert-Butyl Ester, 1,1-Dimethylethyl Cyanoacetate, Cyanoacetic Acid tert-Butyl Ester, NSC 1072, tert-Butyl 2-Cyanoacetate, tert-Butyl Cyanoacetate
InChI Formula InChI=1S/C7H11NO2/c1-7(2,3)10-6(9)4-5-8/h4H2,1-3H3
SMILES CC(C)(C)OC(=O)CC#N
IUPAC Name tert-butyl 2-cyanoacetate
Molecular Weight (g/mol) 141.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.