missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl diethylphosphonoacetate, 95%

Product Code. 10234774 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10234774

Brand: Thermo Scientific Acros 397750010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27784-76-5 | C10H21O5P | 50.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27784-76-5
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 50.23
MDL Number MFCD00075414
InChI Key NFEGNISFSSLEGU-UHFFFAOYSA-N
Synonym tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
PubChem CID 2773685
IUPAC Name tert-butyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC(C)(C)C)OCC

Specifications

Color Colorless
Density 1.0740g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.074
Formula Weight 50.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material tert-Butyl diethylphosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl diethylphosphonoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.