missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl disulfide, 98%

Product Code. 10236040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236040 250 g Glass bottle 250g
10563731 1 kg Glass bottle 1kg
11935731 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236040 Supplier Thermo Scientific Acros Supplier No. 173622500
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 110-06-5 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-06-5
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00008838
InChI Key BKCNDTDWDGQHSD-UHFFFAOYSA-N
Synonym di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane
PubChem CID 66060
IUPAC Name 2-(tert-butyldisulfanyl)-2-methylpropane
SMILES CC(C)(C)SSC(C)(C)C

Specifications

Melting Point -5.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 198.0°C to 204.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2S2
Refractive Index 1.4882 to 1.4902
Quantity 250 g
Beilstein 01,IV,1638
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 178.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl disulfide
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl disulfide
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.