Learn More
tert-Butyl ethyl malonate, 95%
Beskrivning
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.
Solubility
Miscible with most common solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifikationer
Specifikationer
| Density | 0.994 |
| Boiling Point | 98°C to 100°C (22 mmHg) |
| Flash Point | 87°C (188°F) |
| Refractive Index | 1.416 |
| Quantity | 100 g |
| Beilstein | 1776681 |
| Solubility Information | Miscible with most common solvents. Immiscible with water. |
| Formula Weight | 188.22 |
| Percent Purity | 95% |
| Chemical Name or Material | tert-Butyl ethyl malonate |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.