Learn More
tert-Butyl hydrogen malonate, 96%
Descripción
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.
Solubility
Soluble in ethanol.
Notes
Store in a cool, dry condition in a well sealed container.
Especificaciones
Especificaciones
| Melting Point | 19°C to 20°C |
| Density | 1.04 |
| Boiling Point | 90°C (2 mmHg) |
| Flash Point | 91°C (195°F) |
| Refractive Index | 1.426 |
| Quantity | 5 g |
| Beilstein | 1765457 |
| Solubility Information | Soluble in ethanol. |
| Formula Weight | 160.17 |
| Percent Purity | 96% |
| Mostrar más |
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.