missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen malonate, 96%

Product Code. 11346659
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11346659

Brand: Thermo Scientific Alfa Aesar H26223.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40052-13-9 | C7H12O4 | 160.169 g/mol

tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

Solubility
Soluble in ethanol.

Notes
Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40052-13-9
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00191886
InChI Key NGGGZUAEOKRHMA-UHFFFAOYSA-N
Synonym 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
PubChem CID 545853
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
SMILES CC(C)(C)OC(=O)CC(=O)O

Specifications

Melting Point 19°C to 20°C
Density 1.04
Boiling Point 90°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.426
Quantity 25 g
Beilstein 1765457
Solubility Information Soluble in ethanol.
Formula Weight 160.17
Percent Purity 96%
Chemical Name or Material tert-Butyl hydrogen malonate
Show More Show Less

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato