missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl hydroperoxide, 70% Solution in water
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Can
Plastic bottle
Unit Size:
10kg
2.5kg
250g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -2.8°C |
| Color | Colorless |
| Density | 0.9400g/mL |
| Boiling Point | 37.0°C (15.0 mmHg) |
| Flash Point | 43°C |
| Packaging | Plastic bottle |
| Linear Formula | (CH3)3COOH |
| Refractive Index | 1.3860 to 1.3880 |
| Quantity | 2.5 kg |
| Beilstein | 01,IV,1616 |
| Show More |
Safety and Handling
Product Identifier
- tert-Butyl hydroperoxide, 70% solution in water
- Danger
- Carcinogenicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Germ cell mutagenicity Category 2
- Organic peroxide Type E & F
- Skin sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H242-Heating may cause a fire.
- H226-Flammable liquid and vapour.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction